Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(1-Propionylpiperidin-4-yl)-3-(4-(trifluoromethoxy)phenyl)urea 98%

Supplier: AMBEED, INC
Danger

A982864-10MG A982864-1MG A982864-25MG A982864-50MG A982864-5MG
76656-690EA 448.77 USD InStock
76656-690 76656-692 76656-694 76656-696 76656-698 76656-700
1-(1-Propionylpiperidin-4-yl)-3-(4-(trifluoromethoxy)phenyl)urea 98%
1-(1-Propionylpiperidin-4-yl)-3-(4-(trifluoromethoxy)phenyl)urea
Precaución: for Research Use Only


Formula: C₁₆H₂₀F₃N₃O₃
MDL Number: MFCD26142949
CAS Number: 1222780-33-7
UN: 2811
ADR: 6.1,Ⅲ
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty