Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-[(Benzyloxy)methyl]piperidine hydrochloride 95%

Supplier: AMBEED, INC
Warning

A983015-100MG A983015-1G A983015-250MG
76656-702EA 43 USD InStock
76656-702 76656-704 76656-706
4-[(Benzyloxy)methyl]piperidine hydrochloride 95%
4-[(Benzyloxy)methyl]piperidine hydrochloride
Precaución: for Research Use Only


Formula: C₁₃H₂₀ClNO
MW: 241.76 g/mol
MDL Number: MFCD09879195
CAS Number: 301226-92-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty