Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N1,N2-Bis(4-hydroxy-2,6-dimethylphenyl)oxalamide 97%

Supplier: AMBEED, INC
Danger

A983452-1G A983452-250MG A983452-25G A983452-5G
76656-732EA 14 USD InStock
76656-732 76656-734 76656-736 76656-738
N1,N2-Bis(4-hydroxy-2,6-dimethylphenyl)oxalamide 97%
N1,N2-Bis(4-hydroxy-2,6-dimethylphenyl)oxalamide
Precaución: for Research Use Only


Formula: C₁₈H₂₀N₂O₄
MDL Number: MFCD32666963
CAS Number: 1809288-95-6
UN: 3077
ADR: 9,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty