Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Diethyl (4-fluorobenzoyl)malonate 95%

Supplier: AMBEED, INC
Warning

A984615-1G A984615-25G A984615-5G
76656-840EA 10 USD InStock
76656-840 76656-842 76656-844
Diethyl (4-fluorobenzoyl)malonate 95%
Diethyl (4-fluorobenzoyl)malonate
Precaución: for Research Use Only


Formula: C₁₄H₁₅FO₅
MW: 282.2643 g/mol
MDL Number: MFCD29081542
CAS Number: 174403-79-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty