Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(4-Fluorobenzyl)succinic acid 95%

Supplier: AMBEED, INC
Warning

A986022-100MG A986022-250MG A986022-50MG
76656-942EA 35 USD InStock
76656-942 76656-944 77623-282
2-(4-Fluorobenzyl)succinic acid 95%
2-(4-Fluorobenzyl)succinic acid
Precaución: for Research Use Only


Formula: C₁₁H₁₁FO₄
MDL Number: MFCD09835755
CAS Number: 1268121-69-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty