Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5,10,15,20-meso-Tetrakis[4-(methoxycarbonyl)phenyl]porphyrin 96%

Supplier: AMBEED, INC
Warning

A986233-10G A986233-1G A986233-5G
76656-980EA 576 USD InStock
76656-980 76656-982 76656-984
5,10,15,20-meso-Tetrakis[4-(methoxycarbonyl)phenyl]porphyrin 96%
5,10,15,20-meso-Tetrakis[4-(methoxycarbonyl)phenyl]porphyrin
Precaución: for Research Use Only


Formula: C₅₂H₃₈N₄O₈
MDL Number: MFCD00218869
CAS Number: 22112-83-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty