Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Methyl-1-propylpyrrolidinium bis(trifluoromethanesulfonyl)imide 96%

Supplier: AMBEED, INC
Warning

A986356-100G A986356-1G A986356-25G A986356-5G
76656-994EA 288 USD InStock
76656-994 76656-996 76656-998 76657-000
1-Methyl-1-propylpyrrolidinium bis(trifluoromethanesulfonyl)imide 96%
1-Methyl-1-propylpyrrolidinium bis(trifluoromethanesulfonyl)imide
Precaución: for Research Use Only


Formula: C₁₀H₁₈F₆N₂O₄S₂
MDL Number: MFCD08458917
CAS Number: 223437-05-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty