Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Amino-PEG9-acid 95%

Supplier: AMBEED, INC
Warning

A986362-100MG A986362-250MG
76657-002EA 74 USD InStock
76657-002 76657-004
Amino-PEG9-acid 95%
Amino-PEG9-acid
Precaución: for Research Use Only


Formula: C₂₁H₄₃NO₁₁
MDL Number: MFCD26127809
CAS Number: 1191079-83-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty