Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(1,3-Dioxoisoindolin-2-yl)ethyl phosphorodichloridate 95%

Supplier: AMBEED, INC
Danger

A986523-100MG A986523-250MG
76657-012EA 75 USD InStock
76657-012 76657-014
2-(1,3-Dioxoisoindolin-2-yl)ethyl phosphorodichloridate 95%
2-(1,3-Dioxoisoindolin-2-yl)ethyl phosphorodichloridate
Precaución: for Research Use Only


Formula: C₁₀H₈Cl₂NO₄P
MDL Number: MFCD00152165
CAS Number: 52198-45-5
UN: 2811
ADR: 6.1,Ⅲ
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty