Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(Bromomethyl)-4-ethynylbenzene 95%

Supplier: AMBEED, INC
Danger

A987949-100MG A987949-250MG
76657-148EA 94 USD InStock
76657-148 76657-150
1-(Bromomethyl)-4-ethynylbenzene 95%
1-(Bromomethyl)-4-ethynylbenzene
Precaución: For research use only.


Formula: C₉H₇Br
MDL Number: MFCD18968509
CAS Number: 351002-91-0
UN: 3261
ADR: 8,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty