Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(6-Bromobenzo[d]isoxazol-3-yl)acetic acid 95%

Supplier: AMBEED, INC
Warning

A988061-100MG A988061-250MG
76657-156EA 25 USD InStock
76657-156 76657-158
2-(6-Bromobenzo[d]isoxazol-3-yl)acetic acid 95%
2-(6-Bromobenzo[d]isoxazol-3-yl)acetic acid
Precaución: for Research Use Only


Formula: C₉H₆BrNO₃
MDL Number: MFCD27921570
CAS Number: 540750-32-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty