Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Bis(3,5-di-tert-butylphenyl)phosphine oxide 98%

Supplier: AMBEED, INC
Warning

A989624-1G A989624-250MG A989624-5G
76657-286EA 105 USD InStock
76657-286 76657-288 76657-290
Bis(3,5-di-tert-butylphenyl)phosphine oxide 98%
Bis(3,5-di-tert-butylphenyl)phosphine oxide
Precaución: for Research Use Only


Formula: C₂₈H₄₃OP
MW: 426.6142 g/mol
MDL Number: MFCD08458886
CAS Number: 325773-65-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty