Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Chloro-6-methoxy-7-(3-morpholinopropoxy)quinoline-3-carbonitrile 95%

Supplier: AMBEED, INC
Warning

A989882-100MG A989882-1G A989882-250MG
76657-314EA 69 USD InStock
76657-314 76657-316 76657-318
4-Chloro-6-methoxy-7-(3-morpholinopropoxy)quinoline-3-carbonitrile 95%
4-Chloro-6-methoxy-7-(3-morpholinopropoxy)quinoline-3-carbonitrile
Precaución: for Research Use Only


Formula: C₁₈H₂₀ClN₃O₃
MDL Number: MFCD23101039
CAS Number: 214487-30-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty