Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Chloro-2,3-difluorophenylboronic acid pinacol ester 95%

Supplier: AMBEED, INC
Warning

A991801-100MG A991801-1G A991801-250MG
76657-438EA 18 USD InStock
76657-438 76657-440 76657-442
5-Chloro-2,3-difluorophenylboronic acid pinacol ester 95%
5-Chloro-2,3-difluorophenylboronic acid pinacol ester
Precaución: for Research Use Only


Formula: C₁₂H₁₄BClF₂O₂
MDL Number: MFCD29035409
CAS Number: 2246586-13-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty