Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(3-Aminopropyl)-1H-pyrrole-2,5-dione hydrochloride 95%

Supplier: AMBEED, INC
Warning

A992598-100MG A992598-1G A992598-250MG
76657-542EA 27 USD InStock
76657-542 76657-544 76657-546
1-(3-Aminopropyl)-1H-pyrrole-2,5-dione hydrochloride 95%
1-(3-Aminopropyl)-1H-pyrrole-2,5-dione hydrochloride
Precaución: for Research Use Only


Formula: C₇H₁₁ClN₂O₂
MDL Number: MFCD11519188
CAS Number: 1257852-74-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty