Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,5-Dioxopyrrolidin-1-yl heptanoate 95%

Supplier: AMBEED, INC
Warning

A992862-100MG A992862-250MG
76657-582EA 68 USD InStock
76657-582 76657-584
2,5-Dioxopyrrolidin-1-yl heptanoate 95%
2,5-Dioxopyrrolidin-1-yl heptanoate
Precaución: for Research Use Only


Formula: C₁₁H₁₇NO₄
MDL Number: MFCD22193887
CAS Number: 104943-22-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty