Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Imino(4-methoxyphenyl)(methyl)-lambda6-sulfanone 95%

Supplier: AMBEED, INC
Warning

A992911-100MG A992911-250MG
76657-594EA 53 USD InStock
76657-594 76657-596
Imino(4-methoxyphenyl)(methyl)-lambda6-sulfanone 95%
Imino(4-methoxyphenyl)(methyl)-lambda6-sulfanone
Precaución: for Research Use Only


Formula: C₈H₁₁NO₂S
MDL Number: MFCD09878523
CAS Number: 77970-95-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty