Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(tert-Butyl) 2-(4-methoxybenzyl) 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrrole-1,2-dicarboxylate 95%

Supplier: AMBEED, INC
Warning

A993820-1G A993820-250MG A993820-5G
76657-630EA 73 USD InStock
76657-630 76657-632 76657-634
1-(tert-Butyl) 2-(4-methoxybenzyl) 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrrole-1,2-dicarboxylate 95%
1-(tert-Butyl) 2-(4-methoxybenzyl) 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrrole-1,2-dicarboxylate
Precaución: for Research Use Only


Formula: C₂₄H₃₂BNO₇
MDL Number: MFCD29043345
CAS Number: 2377607-06-0
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty