Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Acetyl-1-methyl-1H-pyrazole 95%

Supplier: AMBEED, INC
Warning

A995383-100MG A995383-1G A995383-250MG A995383-5G
76657-702EA 123 USD InStock
76657-702 76657-704 76657-706 76657-708
3-Acetyl-1-methyl-1H-pyrazole 95%
3-Acetyl-1-methyl-1H-pyrazole
Precaución: for Research Use Only


Formula: C₆H₈N₂O
MDL Number: MFCD04968783
CAS Number: 137890-04-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty