Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Methyl-3-propylimidazolium chloride 98% (by HPLC, titration analysis)

Supplier: AMBEED, INC
Warning

A996009-25G A996009-5G
76657-744EA 16 USD InStock
76657-744 76657-746
1-Methyl-3-propylimidazolium chloride 98% (by HPLC, titration analysis)
1-Methyl-3-propylimidazolium chloride
Precaución: for Research Use Only


Formula: C₇H₁₃ClN₂
MDL Number: MFCD17014728
CAS Number: 79917-89-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty