Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Chloro-5-pyrrolidinophenylboronic acid, pinacol ester 96%

Supplier: AMBEED, INC
Warning

A996579-100MG A996579-1G A996579-250MG
76657-756EA 31 USD InStock
76657-756 76657-758 76657-760
3-Chloro-5-pyrrolidinophenylboronic acid, pinacol ester 96%
3-Chloro-5-pyrrolidinophenylboronic acid, pinacol ester
Precaución: for Research Use Only


Formula: C₁₆H₂₃BClNO₂
MDL Number: MFCD18783182
CAS Number: 2096331-84-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty