Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl 16-bromohexadecanoate 97%

Supplier: AMBEED, INC
Warning

A999149-1G A999149-250MG A999149-5G
76657-792EA 25 USD InStock
76657-792 76657-794 76657-796
tert-Butyl 16-bromohexadecanoate 97%
tert-Butyl 16-bromohexadecanoate
Precaución: for Research Use Only


Formula: C₂₀H₃₉BrO₂
MDL Number: MFCD31728047
CAS Number: 865200-89-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty