Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N'-(3-Chloro-4-fluorophenyl)-2-(4-chlorophenyl)-2-oxoacetohydrazonoyl cyanide 98%

Supplier: AMBEED, INC
Warning

A1359457-10MG A1359457-1MG A1359457-5MG
76658-914EA 47 USD InStock
76658-914 76658-916 76658-918
N'-(3-Chloro-4-fluorophenyl)-2-(4-chlorophenyl)-2-oxoacetohydrazonoyl cyanide 98%
N'-(3-Chloro-4-fluorophenyl)-2-(4-chlorophenyl)-2-oxoacetohydrazonoyl cyanide
Precaución: for Research Use Only


Formula: C₁₅H₈Cl₂FN₃O
MDL Number: MFCD00176649
CAS Number: 882290-02-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty