Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Vacuolin-1 98%

Supplier: AMBEED, INC
Warning

A1359469-10MG A1359469-1MG A1359469-5MG
76658-940EA 101 USD InStock
76658-940 76658-942 76658-944
Vacuolin-1 98%
Vacuolin-1
Precaución: for Research Use Only


Formula: C₂₆H₂₄IN₇O
MDL Number: MFCD00321410
CAS Number: 351986-85-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty