Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dibenzyl 2-aminopentanedioate hydrochloride 97%

Supplier: AMBEED, INC
Warning

A1360271-100MG A1360271-1G A1360271-250MG A1360271-5G
76659-000EA 20 USD InStock
76659-000 76659-002 76659-004 76659-006
Dibenzyl 2-aminopentanedioate hydrochloride 97%
Dibenzyl 2-aminopentanedioate hydrochloride
Precaución: for Research Use Only


Formula: C₁₉H₂₂ClNO₄
MW: 363.8353 g/mol
MDL Number: MFCD29037723
CAS Number: 2828432-29-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty