Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2',7'-Dibromospiro[adamantane-2,9'-fluorene] 97%

Supplier: AMBEED, INC
Warning

A1362589-1G A1362589-250MG
76659-148EA 51 USD InStock
76659-148 76659-150
2',7'-Dibromospiro[adamantane-2,9'-fluorene] 97%
2',7'-Dibromospiro[adamantane-2,9'-fluorene]
Precaución: for Research Use Only


Formula: C₂₂H₂₀Br₂
MDL Number: MFCD33395866
CAS Number: 727730-32-7
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty