Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4,4'-Diamino-[1,1'-biphenyl]-3,3'-dicarbonitrile 97%

Supplier: AMBEED, INC
Danger

A1363786-100MG A1363786-1G A1363786-250MG
76659-254EA 56 USD InStock
76659-254 76659-256 77621-778
4,4'-Diamino-[1,1'-biphenyl]-3,3'-dicarbonitrile 97%
4,4'-Diamino-[1,1'-biphenyl]-3,3'-dicarbonitrile
Precaución: for Research Use Only


Formula: C₁₄H₁₀N₄
CAS Number: 61382-01-2
UN: 3439
ADR: 6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty