Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Bromo-5-nitrosalicylaldehyde 95%

Supplier: AMBEED, INC
Danger

A1364120-100G A1364120-5G
76659-296EA 244 USD InStock
76659-296 76659-298
3-Bromo-5-nitrosalicylaldehyde 95%
3-Bromo-5-nitrosalicylaldehyde
Precaución: for Research Use Only


Formula: C₇H₄BrNO₄
MW: 246.02 g/mol
MDL Number: MFCD00051833
CAS Number: 16789-84-7
UN: 2811
ADR: 6.1 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty