Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,6-Dimethoxyphenol 97%, (Chunk or granule or flakes)

Supplier: AMBEED, INC
Warning

A1364713-100G A1364713-5G
76659-368EA 79 USD InStock
76659-368 76659-370
2,6-Dimethoxyphenol 97%, (Chunk or granule or flakes)
2,6-Dimethoxyphenol
Precaución: for Research Use Only


Formula: C₈H₁₀O₃
MDL Number: MFCD00064434
CAS Number: 91-10-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty