Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-Boc-L-Cysteine Methyl ester 90%

Supplier: AMBEED, INC
Danger

A1367188-1G A1367188-25G A1367188-5G
76659-410EA 7 USD InStock
76659-410 76659-412 76659-414
N-Boc-L-Cysteine Methyl ester 90%
N-Boc-L-Cysteine Methyl ester
Precaución: for Research Use Only


Formula: C₉H₁₇NO₄S
MW: 235.3006 g/mol
MDL Number: MFCD00799495
CAS Number: 55757-46-5
UN: 3082
ADR: 9 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty