Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-(tert-Butoxy)-5-oxopentanoic acid ≥98%

Supplier: AMBEED, INC
Danger

A1367256-1G A1367256-25G A1367256-5G A1367256-10G
76659-416EA 13 USD InStock
76659-416 76659-418 76659-420 77621-852
5-(tert-Butoxy)-5-oxopentanoic acid ≥98%
5-(tert-Butoxy)-5-oxopentanoic acid
Precaución: for Research Use Only


Formula: C₉H₁₆O₄
MW: 188.22 g/mol
MDL Number: MFCD09759103
CAS Number: 63128-51-8
UN: 3265
ADR: 8 Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty