Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,1',1'',1'''-(Methanetetrayltetrakis(benzene-4,1-diyl))tetraethanone 97%

Supplier: AMBEED, INC
Warning

A1367272-100MG A1367272-1G A1367272-250MG
76659-422EA 47 USD InStock
76659-422 76659-424 76659-426
1,1',1'',1'''-(Methanetetrayltetrakis(benzene-4,1-diyl))tetraethanone 97%
1,1',1'',1'''-(Methanetetrayltetrakis(benzene-4,1-diyl))tetraethanone
Precaución: for Research Use Only


Formula: C₃₃H₂₈O₄
MDL Number: MFCD30531760
CAS Number: 313484-93-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty