Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,4-Dichloro-N-(5-chloro-2-(hydrazinylmethyl)phenyl)benzenesulfonamide hydrochloride 97%

Supplier: AMBEED, INC
Warning

A1367274-100MG A1367274-250MG
76659-428EA 112 USD InStock
76659-428 76659-430
3,4-Dichloro-N-(5-chloro-2-(hydrazinylmethyl)phenyl)benzenesulfonamide hydrochloride 97%
3,4-Dichloro-N-(5-chloro-2-(hydrazinylmethyl)phenyl)benzenesulfonamide hydrochloride
Precaución: for Research Use Only


Formula: C₁₃H₁₃Cl₄N₃O₂S
MW: 417.1382 g/mol
MDL Number: MFCD33022451
CAS Number: 2682114-55-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty