Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

trans-4-Chloro-β-nitrostyrene 98% predominantly trans

Supplier: AMBEED, INC
Danger

A1369280-1G A1369280-5G A1369280-25G
76659-510EA 9 USD InStock
76659-510 76659-512 77621-860
trans-4-Chloro-β-nitrostyrene 98% predominantly trans
trans-4-Chloro-β-nitrostyrene
Precaución: for Research Use Only


Formula: C₈H₆ClNO₂
MDL Number: MFCD00024825
CAS Number: 706-07-0
UN: 3077
ADR: 9,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty