Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(2,2,2-Trifluoroethoxy)pyrrolidine hydrochloride 97%

Supplier: AMBEED, INC
Warning

A1369582-100MG A1369582-1G A1369582-250MG A1369582-50MG
76659-526EA 109 USD InStock
76659-526 76659-528 76659-530 76659-532
3-(2,2,2-Trifluoroethoxy)pyrrolidine hydrochloride 97%
3-(2,2,2-Trifluoroethoxy)pyrrolidine hydrochloride
Precaución: for Research Use Only


Formula: C₆H₁₁ClF₃NO
MDL Number: MFCD34178736
CAS Number: 2703752-17-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty