Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Bromo-7-fluorobenzo[d]oxazole 97%

Supplier: AMBEED, INC
Warning

A1371082-100MG A1371082-1G A1371082-250MG
76659-652EA 204 USD InStock
76659-652 76659-654 76659-656
6-Bromo-7-fluorobenzo[d]oxazole 97%
6-Bromo-7-fluorobenzo[d]oxazole
Precaución: for Research Use Only


Formula: C₇H₃BrFNO
MDL Number: MFCD27664915
CAS Number: 1823959-85-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty