Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,3'-Dibromo-4,4'-biphenol 98%

Supplier: AMBEED, INC
Warning

A1371977-100MG A1371977-1G A1371977-250MG
76659-818EA 9 USD InStock
76659-818 76659-820 76659-822
3,3'-Dibromo-4,4'-biphenol 98%
3,3'-Dibromo-4,4'-biphenol
Precaución: for Research Use Only


Formula: C₁₂H₈Br₂O₂
MW: 344.00 g/mol
MDL Number: MFCD10566892
CAS Number: 189039-64-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty