Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

trans-2-Dodecenal 10% triethyl citrate

Supplier: AMBEED, INC
Warning

A1372192-100G A1372192-25G A1372192-5G
76659-854EA 155 USD InStock
76659-854 76659-856 76659-858
trans-2-Dodecenal 10% triethyl citrate
trans-2-Dodecenal
Precaución: for Research Use Only


Formula: C₁₂H₂₂O
MW: 182.31 g/mol
MDL Number: MFCD00014674
CAS Number: 20407-84-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty