Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Diethyl 2-acetamido-2-(2-chloro-4-methylbenzyl)malonate 97%

Supplier: AMBEED, INC
Warning

A1372344-100MG A1372344-1G A1372344-250MG A1372344-5G
76659-866EA 15 USD InStock
76659-866 76659-868 76659-870 76659-872
Diethyl 2-acetamido-2-(2-chloro-4-methylbenzyl)malonate 97%
Diethyl 2-acetamido-2-(2-chloro-4-methylbenzyl)malonate
Precaución: for Research Use Only


Formula: C₁₇H₂₂ClNO₅
MW: 355.8133 g/mol
MDL Number: MFCD33022471
CAS Number: 2682114-30-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty