Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

ɑ-Naphthol orange BS

Supplier: AMBEED, INC
Warning

A1372858-100G A1372858-25G A1372858-500G
76659-958EA 26 USD InStock
76659-958 76659-960 76659-962
ɑ-Naphthol orange BS
alpha-Naphthol orange
Precaución: for Research Use Only


Formula: C₁₆H₁₂N₂O₄S
MW: 350.32 g/mol
MDL Number: MFCD00021510
CAS Number: 523-44-4
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty