Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N1-(Furan-2-ylmethyl)-N2-(naphthalen-1-yl)oxalamide 97%

Supplier: AMBEED, INC
Danger

A1373480-100MG A1373480-1G A1373480-250MG A1373480-5G
76660-034EA 36 USD InStock
76660-034 76660-036 76660-038 76660-040
N1-(Furan-2-ylmethyl)-N2-(naphthalen-1-yl)oxalamide 97%
N1-(Furan-2-ylmethyl)-N2-(naphthalen-1-yl)oxalamide
Precaución: for Research Use Only


Formula: C₁₇H₁₄N₂O₃
MDL Number: MFCD01987779
CAS Number: 428840-15-7
UN: 3077
ADR: 9,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty