Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(Piperazin-1-yl)isonicotinic acid hydrochloride 97%

Supplier: AMBEED, INC
Warning

A1373566-100MG A1373566-250MG
76660-054EA 34 USD InStock
76660-054 76660-056
2-(Piperazin-1-yl)isonicotinic acid hydrochloride 97%
2-(Piperazin-1-yl)isonicotinic acid hydrochloride
Precaución: for Research Use Only


Formula: C₁₀H₁₄ClN₃O₂
MW: 243.6901 g/mol
MDL Number: MFCD33022476
CAS Number: 2682114-49-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty