Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ruthenium(2+), tris(2,2'-bipyrimidine-κN1,κN1')-, (OC-6-11)-, hexafluorophosphate(1-) (1:2) 97%

Supplier: AMBEED, INC
Warning

A1374157-100MG A1374157-1G A1374157-250MG A1374157-50MG
76660-086EA 47 USD InStock
76660-086 76660-088 76660-090 76660-092
Ruthenium(2+), tris(2,2'-bipyrimidine-κN1,κN1')-, (OC-6-11)-, hexafluorophosphate(1-) (1:2) 97%
Ruthenium(2+), tris(2,2'-bipyrimidine-κN1,κN1')-, (OC-6-11)-, hexafluorophosphate(1-) (1:2)
Precaución: for Research Use Only


Formula: C₂₄H₁₈F₁₂N₁₂P₂Ru
CAS Number: 85335-55-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty