Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Iodopentan-1-ol 98% stabilised

Supplier: AMBEED, INC
Danger

Stabilization: Stabilised with copper chip

A1376694-100MG A1376694-250MG A1376694-5G
76660-264EA 13 USD InStock
76660-264 76660-266 77621-960
5-Iodopentan-1-ol 98% stabilised
5-Iodopentan-1-ol
Precaución: for Research Use Only


Formula: C₅H₁₁IO
MW: 214.0447 g/mol
MDL Number: MFCD20415154
CAS Number: 67133-88-4
UN: 2810
ADR: 6.1 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty