Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sodium 2-aminobenzo[d]thiazole-5-carboxylate 97%

Supplier: AMBEED, INC
Warning

A1376706-100MG A1376706-1G A1376706-250MG
76660-270EA 125 USD InStock
76660-270 76660-272 76660-274
Sodium 2-aminobenzo[d]thiazole-5-carboxylate 97%
Sodium 2-aminobenzo[d]thiazole-5-carboxylate
Precaución: for Research Use Only


Formula: C₈H₅N₂NaO₂S
MW: 216.19 g/mol
MDL Number: MFCD34178742
CAS Number: 2703756-61-8
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty