Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Polyethylene glycol 12-hydroxystearate Hyroxyl Value (90-110mg); Acid Value (0.3mg)

Supplier: AMBEED, INC
Warning

A1378061-5G A1378061-100G
76660-294EA 18 USD InStock
76660-294 77597-080
Polyethylene glycol 12-hydroxystearate Hyroxyl Value (90-110mg); Acid Value (0.3mg)
Polyethylene glycol 12-hydroxystearate
Precaución: for Research Use Only


MDL Number: MFCD32661415
CAS Number: 61909-81-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty