Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(2-Chloroethyl)-1H-pyrrolo[2,3-b]pyridine 98%

Supplier: AMBEED, INC
Danger

A1382367-100MG A1382367-1G A1382367-250MG
76660-450EA 36 USD InStock
76660-450 76660-452 76660-454
3-(2-Chloroethyl)-1H-pyrrolo[2,3-b]pyridine 98%
3-(2-Chloroethyl)-1H-pyrrolo[2,3-b]pyridine
Precaución: for Research Use Only


Formula: C₉H₉ClN₂
MDL Number: MFCD20623338
CAS Number: 90929-74-1
UN: 2811
ADR: 6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty