Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(4-Fluorophenyl)-1,3,2-dioxaborinane 98%

Supplier: AMBEED, INC
Warning

A1414182-1G A1414182-250MG A1414182-5G
76660-616EA 25 USD InStock
76660-616 76660-618 76660-620
2-(4-Fluorophenyl)-1,3,2-dioxaborinane 98%
2-(4-Fluorophenyl)-1,3,2-dioxaborinane
Precaución: for Research Use Only


Formula: C₉H₁₀BFO₂
CAS Number: 156942-21-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty