Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Phenylisobutyric acid 98%

Supplier: AMBEED, INC
Warning

A141899-1000G A141899-100G A141899-10G A141899-25G A141899-500G
76660-684EA 156 USD InStock
76660-684 76660-686 76660-688 76660-690 76660-692
2-Phenylisobutyric acid 98%
2-Phenylisobutyric acid
Precaución: for Research Use Only


Formula: C₁₀H₁₂O₂
MDL Number: MFCD00014332
CAS Number: 826-55-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty