Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-3-fluorophthalic acid 97%

Supplier: AMBEED, INC
Warning

A1441401-100MG A1441401-1G A1441401-250MG A1441401-50MG
76660-994EA 26 USD InStock
76660-994 76660-996 76660-998 76661-000
4-Bromo-3-fluorophthalic acid 97%
4-Bromo-3-fluorophthalic acid
Precaución: for Research Use Only


Formula: C₈H₄BrFO₄
CAS Number: 2408642-17-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty